missing translation for 'onlineSavingsMsg'
Learn More

5-Bromovanillin 95.0+%, TCI America™

Catalog No. p-7118898
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11725G 5 g
B117225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B11725G Supplier TCI America Supplier No. B11725G

Chemical Identifiers

CAS 2973-76-4
Molecular Formula C8H7BrO3
Molecular Weight (g/mol) 231.05
MDL Number MFCD00006940
InChI Key KLSHZDPXXKAHIJ-UHFFFAOYSA-N
Synonym 5-bromovanillin, benzaldehyde, 3-bromo-4-hydroxy-5-methoxy, vanillin, 5-bromo, 5-bromo-4-hydroxy-3-methoxybenzaldehyde, 3-bromo-4-hydroxy-5-methoxy-benzaldehyde, unii-508wvs56pw, 3-bromo-4-hydroxy-5-methyloxy benzaldehyde, 5-bromovaniline, 5-bromovanilline
PubChem CID 18099
IUPAC Name 3-bromo-4-hydroxy-5-methoxybenzaldehyde
SMILES COC1=CC(C=O)=CC(Br)=C1O

Specifications

Melting Point 161°C
Quantity 5 g
Formula Weight 231.05
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromovanillin