missing translation for 'onlineSavingsMsg'
Learn More

5-Bromovaleric acid, 97%

Catalog No. p-3777733
Change view
Click to view available options
Quantity:
50 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC169820500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC169820500 Supplier Thermo Scientific Chemicals Supplier No. 169820500

CAS: 2067-33-6 | C5H9BrO2 | 181.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2067-33-6
Molecular Formula C5H9BrO2
Molecular Weight (g/mol) 181.03
MDL Number MFCD00004414
InChI Key WNXNUPJZWYOKMW-UHFFFAOYSA-N
Synonym 5-bromovaleric acid, pentanoic acid, 5-bromo, 5-bromo-n-valeric acid, valeric acid, 5-bromo, 5-bromovalenc acid, 1-bromo-4-carboxybutane, .delta.-bromovaleric acid, delta-bromovaleric acid, 5-bromo valeric acid, 5-bromoval eric acid
PubChem CID 16368
IUPAC Name 5-bromopentanoic acid
SMILES C(CCBr)CC(=O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 37°C to 41°C
Color Yellow to Brown
Boiling Point 142°C to 145°C (13.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula Br(CH2)4CO2H
Quantity 50 g
Beilstein 02, 303
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol
Formula Weight 181.03
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 5-Bromovaleric acid
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 218-185-3

RUO – Research Use Only