Learn More
5-Bromovaleric acid, 97%
CAS: 2067-33-6 | C5H9BrO2 | 181.029 g/mol
Supplier: Thermo Scientific Chemicals A1176522
| Quantity | 100 g |
|---|
Alert:
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Description
5-Bromovaleric acid is used in the synthesis of inhibitors for aminoglycoside resistant bacteria. 5-Bromovaleric acid is also used in the preparation of sequence specific base pair mimics as topoisomerase IB inhibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Bromovaleric acid is used in the synthesis of inhibitors for aminoglycoside resistant bacteria. 5-Bromovaleric acid is also used in the preparation of sequence specific base pair mimics as topoisomerase IB inhibitors.
Solubility
Soluble in ethanol. Insoluble in water.
Notes
Store away from strong bases and oxidizing agents. Store at room temperature.
Chemical Identifiers
| C5H9BrO2 | |
| MFCD00004414 | |
| 5-bromovaleric acid, pentanoic acid, 5-bromo, 5-bromo-n-valeric acid, valeric acid, 5-bromo, 5-bromovalenc acid, 1-bromo-4-carboxybutane, .delta.-bromovaleric acid, delta-bromovaleric acid, 5-bromo valeric acid, 5-bromoval eric acid | |
| 5-bromopentanoic acid |
Specifications
| 5-Bromovaleric acid | |
| 38°C to 41°C | |
| >100°C (212°F) | |
| 100 g | |
| UN3261 | |
| 5-bromovaleric acid, pentanoic acid, 5-bromo, 5-bromo-n-valeric acid, valeric acid, 5-bromo, 5-bromovalenc acid, 1-bromo-4-carboxybutane, .delta.-bromovaleric acid, delta-bromovaleric acid, 5-bromo valeric acid, 5-bromoval eric acid | |
| WNXNUPJZWYOKMW-UHFFFAOYSA-N | |
| 5-bromopentanoic acid | |
| 16368 | |
| 97% |
| 2067-33-6 | |
| 142°C to 145°C (13 mmHg) | |
| C5H9BrO2 | |
| MFCD00004414 | |
| 1745664 | |
| Soluble in ethanol. Insoluble in water. | |
| C(CCBr)CC(=O)O | |
| 181.029 | |
| 181.03 |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 218-185-3
TSCA : Yes
Recommended Storage : Ambient temperatures