missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromouridine 99.0+%, TCI America™
$34.66 - $134.75
Chemical Identifiers
| CAS | 957-75-5 |
|---|---|
| Molecular Formula | C9H11BrN2O6 |
| Molecular Weight (g/mol) | 323.10 |
| MDL Number | MFCD00006528 |
| InChI Key | AGFIRQJZCNVMCW-UHFFFAOYNA-N |
| Synonym | 5-bromouridine, uridine, 5-bromo, 1-beta-ribofuranosyl-5-bromo-uracil, 5-bromo-1-2r,3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidine-2,4 1h,3h-dione, bromouridine, 5-bromo-uridine, key8pg1brc, unii-key8pg1brc, 5-bromouracil ribonucleoside, bru |
| PubChem CID | 91494 |
| ChEBI | CHEBI:20553 |
| IUPAC Name | 5-bromo-1-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione |
| SMILES | OCC1OC(C(O)C1O)N1C=C(Br)C(=O)NC1=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B0666100MG
|
TCI America
B0666100MG |
100 mg |
Each for $34.66
|
|
|||||
|
B06661G
|
TCI America
B06661G |
1 g |
Each for $134.75
|
|
|||||
Chemical Identifiers
| 957-75-5 | |
| 323.10 | |
| AGFIRQJZCNVMCW-UHFFFAOYNA-N | |
| 91494 | |
| 5-bromo-1-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione |
| C9H11BrN2O6 | |
| MFCD00006528 | |
| 5-bromouridine, uridine, 5-bromo, 1-beta-ribofuranosyl-5-bromo-uracil, 5-bromo-1-2r,3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidine-2,4 1h,3h-dione, bromouridine, 5-bromo-uridine, key8pg1brc, unii-key8pg1brc, 5-bromouracil ribonucleoside, bru | |
| CHEBI:20553 | |
| OCC1OC(C(O)C1O)N1C=C(Br)C(=O)NC1=O |
Specifications
| 957-75-5 | |
| White | |
| MFCD00006528 | |
| 5-bromouridine, uridine, 5-bromo, 1-beta-ribofuranosyl-5-bromo-uracil, 5-bromo-1-2r,3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidine-2,4 1h,3h-dione, bromouridine, 5-bromo-uridine, key8pg1brc, unii-key8pg1brc, 5-bromouracil ribonucleoside, bru | |
| OCC1OC(C(O)C1O)N1C=C(Br)C(=O)NC1=O | |
| 323.10 | |
| CHEBI:20553 | |
| ≥99.0% (HPLC) | |
| 5-Bromouridine |
| 182°C | |
| C9H11BrN2O6 | |
| 100 mg | |
| AGFIRQJZCNVMCW-UHFFFAOYNA-N | |
| 5-bromo-1-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione | |
| 91494 | |
| 323.10 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : YU7300000
TSCA : No
Recommended Storage : Refrigerator