missing translation for 'onlineSavingsMsg'
Learn More

5-Bromouridine 99.0+%, TCI America™

Catalog No. B0666100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0666100MG 100 mg
B06661G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0666100MG Supplier TCI America Supplier No. B0666100MG

Chemical Identifiers

CAS 957-75-5
Molecular Formula C9H11BrN2O6
Molecular Weight (g/mol) 323.10
MDL Number MFCD00006528
InChI Key AGFIRQJZCNVMCW-UHFFFAOYNA-N
Synonym 5-bromouridine, uridine, 5-bromo, 1-beta-ribofuranosyl-5-bromo-uracil, 5-bromo-1-2r,3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidine-2,4 1h,3h-dione, bromouridine, 5-bromo-uridine, key8pg1brc, unii-key8pg1brc, 5-bromouracil ribonucleoside, bru
PubChem CID 91494
ChEBI CHEBI:20553
IUPAC Name 5-bromo-1-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES OCC1OC(C(O)C1O)N1C=C(Br)C(=O)NC1=O

Specifications

Melting Point 182°C
Color White
Quantity 100 mg
Formula Weight 323.10
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromouridine
RTECSNumber YU7300000
TSCA No
Recommended Storage Refrigerator