missing translation for 'onlineSavingsMsg'
Learn More

5-Bromouracil 98.0+%, TCI America™

Supplier:  TCI America B066525G

Catalog No. B066525G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

5-Bromouracil
C4H3BrN2O2
25 g
LQLQRFGHAALLLE-UHFFFAOYSA-N
5-bromo-1,2,3,4-tetrahydropyrimidine-2,4-dione
5802
190.98
Crystalline Powder
51-20-7
MFCD00006017
5-bromouracil, bromouracil, uracil, 5-bromo, 5-bromopyrimidine-2,4 1h,3h-dione, 5-bromo-2,4 1h,3h-pyrimidinedione, 5-bromo-2,4-dihydroxypyrimidine, 2,4 1h,3h-pyrimidinedione, 5-bromo, 5-bromopyrimidine-2,4-diol, unii-4hk400g5uo, 5-bromo-1,2,3,4-tetrahydropyrimidine-2,4-dione
BrC1=CNC(=O)NC1=O
190.98
CHEBI:20552
≥98.0% (T)

Chemical Identifiers

51-20-7
190.98
LQLQRFGHAALLLE-UHFFFAOYSA-N
5802
5-bromo-1,2,3,4-tetrahydropyrimidine-2,4-dione
C4H3BrN2O2
MFCD00006017
5-bromouracil, bromouracil, uracil, 5-bromo, 5-bromopyrimidine-2,4 1h,3h-dione, 5-bromo-2,4 1h,3h-pyrimidinedione, 5-bromo-2,4-dihydroxypyrimidine, 2,4 1h,3h-pyrimidinedione, 5-bromo, 5-bromopyrimidine-2,4-diol, unii-4hk400g5uo, 5-bromo-1,2,3,4-tetrahydropyrimidine-2,4-dione
CHEBI:20552
BrC1=CNC(=O)NC1=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : YQ9060000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions