missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 5-Bromouracil, 98%

CAS: 51-20-7 | C4H3BrN2O2 | 190.98 g/mol

$51.10 - $51.10

Chemical Identifiers

CAS 51-20-7
Molecular Formula C4H3BrN2O2
Molecular Weight (g/mol) 190.98
MDL Number MFCD00006017
InChI Key LQLQRFGHAALLLE-UHFFFAOYSA-N
Synonym 5-bromouracil, bromouracil, uracil, 5-bromo, 5-bromopyrimidine-2,4 1h,3h-dione, 5-bromo-2,4 1h,3h-pyrimidinedione, 5-bromo-2,4-dihydroxypyrimidine, 2,4 1h,3h-pyrimidinedione, 5-bromo, 5-bromopyrimidine-2,4-diol, unii-4hk400g5uo, 5-bromo-1,2,3,4-tetrahydropyrimidine-2,4-dione
PubChem CID 5802
ChEBI CHEBI:20552
IUPAC Name 5-bromo-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES BrC1=CNC(=O)NC1=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC226440050
SDS View Documents
Thermo Scientific Chemicals
226440050
5 g Glass bottle
Each for $51.10
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

51-20-7
190.98
LQLQRFGHAALLLE-UHFFFAOYSA-N
5802
5-bromo-1,2,3,4-tetrahydropyrimidine-2,4-dione
C4H3BrN2O2
MFCD00006017
5-bromouracil, bromouracil, uracil, 5-bromo, 5-bromopyrimidine-2,4 1h,3h-dione, 5-bromo-2,4 1h,3h-pyrimidinedione, 5-bromo-2,4-dihydroxypyrimidine, 2,4 1h,3h-pyrimidinedione, 5-bromo, 5-bromopyrimidine-2,4-diol, unii-4hk400g5uo, 5-bromo-1,2,3,4-tetrahydropyrimidine-2,4-dione
CHEBI:20552
BrC1=CNC(=O)NC1=O

Specifications

51-20-7
100.0
97.5% min. (HPLC)
C4H3BrN2O2
5 g
15,145
Solubility in water: soluble in cold water. Other solubilities: soluble in ammonia water and other alkalines
BrC1=CNC(=O)NC1=O
190.98
CHEBI:20552
98%
97.5
307.0°C to 310.0°C
Glass bottle
MFCD00006017
24,318
5-bromouracil, bromouracil, uracil, 5-bromo, 5-bromopyrimidine-2,4 1h,3h-dione, 5-bromo-2,4 1h,3h-pyrimidinedione, 5-bromo-2,4-dihydroxypyrimidine, 2,4 1h,3h-pyrimidinedione, 5-bromo, 5-bromopyrimidine-2,4-diol, unii-4hk400g5uo, 5-bromo-1,2,3,4-tetrahydropyrimidine-2,4-dione
LQLQRFGHAALLLE-UHFFFAOYSA-N
5-bromo-1,2,3,4-tetrahydropyrimidine-2,4-dione
5802
190.98
5-Bromouracil, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove

GHS Signal Word: Warning

EINECSNumber : 200-084-0

RTECSNumber : YQ9060000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only