Learn More
5-Bromothiophene-2-sulfonyl chloride, 96%
CAS: 55854-46-1 | C4H2BrClO2S2 | 261.53 g/mol
$187.12 - $640.81
Chemical Identifiers
| CAS | 55854-46-1 |
|---|---|
| Molecular Formula | C4H2BrClO2S2 |
| Molecular Weight (g/mol) | 261.53 |
| MDL Number | MFCD00084909 |
| InChI Key | WGYBIEOLAFYDEC-UHFFFAOYSA-N |
| Synonym | 5-bromothiophenesulfonyl chloride, 5-bromo-2-thiophenesulfonyl chloride, 5-bromo-thiophene-2-sulfonyl chloride, 5-bromothiophene-2-sulphonyl chloride, 5-bromo-2-chlorosulphonyl thiophene, 5-bromo-2-chlorosulfonyl thiophene, 5-bromothiophenesulphonyl chloride, buttpark 21\07-09, 5-bromo 2-thienyl chlorosulfone, 5-bromothiophen-2-sulfonyl chloride |
| PubChem CID | 2733924 |
| IUPAC Name | 5-bromothiophene-2-sulfonyl chloride |
| SMILES | ClS(=O)(=O)C1=CC=C(Br)S1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1467706
|
Thermo Scientific Chemicals
A1467706 |
5 g |
Each for $187.12
|
|
|||||
|
AAA1467714
|
Thermo Scientific Chemicals
A1467714 |
25 g |
Each for $640.81
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 55854-46-1 | |
| 261.53 | |
| WGYBIEOLAFYDEC-UHFFFAOYSA-N | |
| 2733924 | |
| ClS(=O)(=O)C1=CC=C(Br)S1 |
| C4H2BrClO2S2 | |
| MFCD00084909 | |
| 5-bromothiophenesulfonyl chloride, 5-bromo-2-thiophenesulfonyl chloride, 5-bromo-thiophene-2-sulfonyl chloride, 5-bromothiophene-2-sulphonyl chloride, 5-bromo-2-chlorosulphonyl thiophene, 5-bromo-2-chlorosulfonyl thiophene, 5-bromothiophenesulphonyl chloride, buttpark 21\07-09, 5-bromo 2-thienyl chlorosulfone, 5-bromothiophen-2-sulfonyl chloride | |
| 5-bromothiophene-2-sulfonyl chloride |
Specifications
| 55854-46-1 | |
| 99°C to 101°C (0.5 mmHg) | |
| MFCD00084909 | |
| UN3261 | |
| 5-bromothiophenesulfonyl chloride, 5-bromo-2-thiophenesulfonyl chloride, 5-bromo-thiophene-2-sulfonyl chloride, 5-bromothiophene-2-sulphonyl chloride, 5-bromo-2-chlorosulphonyl thiophene, 5-bromo-2-chlorosulfonyl thiophene, 5-bromothiophenesulphonyl chloride, buttpark 21\07-09, 5-bromo 2-thienyl chlorosulfone, 5-bromothiophen-2-sulfonyl chloride | |
| ClS(=O)(=O)C1=CC=C(Br)S1 | |
| 261.53 | |
| 261.55 | |
| 5-Bromothiophene-2-sulfonyl chloride |
| 39°C to 42°C | |
| C4H2BrClO2S2 | |
| 5 g | |
| Moisture sensitive | |
| WGYBIEOLAFYDEC-UHFFFAOYSA-N | |
| 5-bromothiophene-2-sulfonyl chloride | |
| 2733924 | |
| 98% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Hazard Class: 8; Packaging Group: III
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only