missing translation for 'onlineSavingsMsg'
Learn More

5-Bromothiophene-2-sulfonyl chloride, 96%

Catalog No. AAA1467706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1467706 5 g
AAA1467714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1467706 Supplier Thermo Scientific Chemicals Supplier No. A1467706

CAS: 55854-46-1 | C4H2BrClO2S2 | 261.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 55854-46-1
Molecular Formula C4H2BrClO2S2
Molecular Weight (g/mol) 261.53
MDL Number MFCD00084909
InChI Key WGYBIEOLAFYDEC-UHFFFAOYSA-N
Synonym 5-bromothiophenesulfonyl chloride, 5-bromo-2-thiophenesulfonyl chloride, 5-bromo-thiophene-2-sulfonyl chloride, 5-bromothiophene-2-sulphonyl chloride, 5-bromo-2-chlorosulphonyl thiophene, 5-bromo-2-chlorosulfonyl thiophene, 5-bromothiophenesulphonyl chloride, buttpark 21\07-09, 5-bromo 2-thienyl chlorosulfone, 5-bromothiophen-2-sulfonyl chloride
PubChem CID 2733924
IUPAC Name 5-bromothiophene-2-sulfonyl chloride
SMILES ClS(=O)(=O)C1=CC=C(Br)S1

Specifications

Melting Point 39°C to 42°C
Boiling Point 99°C to 101°C (0.5 mmHg)
Quantity 5 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 261.55
Percent Purity 98%
Chemical Name or Material 5-Bromothiophene-2-sulfonyl chloride
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only