missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromosalicylaldehyde 98.0+%, TCI America™
$171.49 - $171.49
Chemical Identifiers
| CAS | 1761-61-1 |
|---|---|
| Molecular Formula | C7H5BrO2 |
| Molecular Weight (g/mol) | 201.019 |
| MDL Number | MFCD00003330 |
| InChI Key | MKKSTJKBKNCMRV-UHFFFAOYSA-N |
| Synonym | 5-bromosalicylaldehyde, benzaldehyde, 5-bromo-2-hydroxy, 2-hydroxy-5-bromobenzaldehyde, salicylaldehyde, 5-bromo, 5-bromo-salicyclaldehyde, unii-0z65a0a8ce, 5-bromo-2-hydroxy-benzaldehyde, 5-bromo-salicyclic aldehyde, 5-bromo-2-hydroxy benzaldehyde, pubchem3779 |
| PubChem CID | 72863 |
| IUPAC Name | 5-bromo-2-hydroxybenzaldehyde |
| SMILES | C1=CC(=C(C=C1Br)C=O)O |
Chemical Identifiers
| 1761-61-1 | |
| 201.019 | |
| MKKSTJKBKNCMRV-UHFFFAOYSA-N | |
| 72863 | |
| C1=CC(=C(C=C1Br)C=O)O |
| C7H5BrO2 | |
| MFCD00003330 | |
| 5-bromosalicylaldehyde, benzaldehyde, 5-bromo-2-hydroxy, 2-hydroxy-5-bromobenzaldehyde, salicylaldehyde, 5-bromo, 5-bromo-salicyclaldehyde, unii-0z65a0a8ce, 5-bromo-2-hydroxy-benzaldehyde, 5-bromo-salicyclic aldehyde, 5-bromo-2-hydroxy benzaldehyde, pubchem3779 | |
| 5-bromo-2-hydroxybenzaldehyde |
Specifications
| 1761-61-1 | |
| White-Yellow | |
| MFCD00003330 | |
| 5-bromosalicylaldehyde, benzaldehyde, 5-bromo-2-hydroxy, 2-hydroxy-5-bromobenzaldehyde, salicylaldehyde, 5-bromo, 5-bromo-salicyclaldehyde, unii-0z65a0a8ce, 5-bromo-2-hydroxy-benzaldehyde, 5-bromo-salicyclic aldehyde, 5-bromo-2-hydroxy benzaldehyde, pubchem3779 | |
| C1=CC(=C(C=C1Br)C=O)O | |
| 201.019 | |
| 201.02 | |
| Crystalline Powder |
| 105°C | |
| C7H5BrO2 | |
| 25 g | |
| MKKSTJKBKNCMRV-UHFFFAOYSA-N | |
| 5-bromo-2-hydroxybenzaldehyde | |
| 72863 | |
| ≥98.0% (GC,T) | |
| 5-Bromosalicylaldehyde |
Safety and Handling
TSCA : Yes