missing translation for 'onlineSavingsMsg'
Learn More

5-Bromosalicylaldehyde, 98%

Catalog No. aaa1191722
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
500 g
Catalog No. Quantity
AAA1191722 100 g
AAA1191714 25 g
AAA1191736 500 g
3 options

Catalog No. AAA1191722

Supplier: Thermo Scientific Chemicals A1191722

Only null left

CAS: 1761-61-1 | C7H5BrO2 | 201.019 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1761-61-1
Molecular Formula C7H5BrO2
Molecular Weight (g/mol) 201.019
MDL Number MFCD00003330
InChI Key MKKSTJKBKNCMRV-UHFFFAOYSA-N
Synonym 5-bromosalicylaldehyde, benzaldehyde, 5-bromo-2-hydroxy, 2-hydroxy-5-bromobenzaldehyde, salicylaldehyde, 5-bromo, 5-bromo-salicyclaldehyde, unii-0z65a0a8ce, 5-bromo-2-hydroxy-benzaldehyde, 5-bromo-salicyclic aldehyde, 5-bromo-2-hydroxy benzaldehyde, pubchem3779
PubChem CID 72863
IUPAC Name 5-bromo-2-hydroxybenzaldehyde
SMILES C1=CC(=C(C=C1Br)C=O)O

Specifications

Melting Point 103°C to 106°C
Quantity 100 g
Beilstein 774710
Sensitivity Air Sensitive
Formula Weight 201.02
Percent Purity 98%
Chemical Name or Material 5-Bromosalicylaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 217-167-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only