missing translation for 'onlineSavingsMsg'
Learn More

5-Bromopyrimidine 98.0+%, TCI America™

Catalog No. p-7119566
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
B22295G 5 g
B222925G 25 g
2 options

Catalog No. B22295G

Supplier: TCI America B22295G

Only null left

Chemical Identifiers

CAS 4595-59-9
Molecular Formula C4H3BrN2
Molecular Weight (g/mol) 158.986
MDL Number MFCD00006117
InChI Key GYCPLYCTMDTEPU-UHFFFAOYSA-N
Synonym pyrimidine, 5-bromo, 5-bromo pyrimidine, 5-bromo-1,3-diazine, 5-bromo-pyrimidine, 5-brpy compound, 5-brompyrimidin, 5-brornopyrimidine, zlchem 349, pubchem6902, 11-cis-retinyl-palmitate
PubChem CID 78344
IUPAC Name 5-bromopyrimidine
SMILES C1=C(C=NC=N1)Br

Specifications

Melting Point 74°C
Color White-Yellow
Boiling Point 168°C
Quantity 5 g
Formula Weight 158.99
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromopyrimidine