missing translation for 'onlineSavingsMsg'
Learn More

5-Bromopyrimidine, 98%

Catalog No. AAL1417222
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1417222 100 g
AAL1417206 5 g
AAL1417214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1417222 Supplier Thermo Scientific Chemicals Supplier No. L1417222
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4595-59-9 | C4H3BrN2 | 158.986 g/mol

5-Bromopyrimidine was used in the synthesis of N-heteroaryl substituted 9-arylcarbazolyl derivatives via palladium-catalyzed aerobic and ligand-free Suzuki reaction, (5-(phenylethynyl)pyrimidine) via microwave assisted organic synthesis (MAOS) Sonogashira protocol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromopyrimidine was used in the synthesis of N-heteroaryl substituted 9-arylcarbazolyl derivatives via palladium-catalyzed aerobic and ligand-free Suzuki reaction, (5-(phenylethynyl)pyrimidine) via microwave assisted organic synthesis (MAOS) Sonogashira protocol.

Solubility
Soluble in chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.

Chemical Identifiers

CAS 4595-59-9
Molecular Formula C4H3BrN2
Molecular Weight (g/mol) 158.986
MDL Number MFCD00006117
InChI Key GYCPLYCTMDTEPU-UHFFFAOYSA-N
Synonym pyrimidine, 5-bromo, 5-bromo pyrimidine, 5-bromo-1,3-diazine, 5-bromo-pyrimidine, 5-brpy compound, 5-brompyrimidin, 5-brornopyrimidine, zlchem 349, pubchem6902, 11-cis-retinyl-palmitate
PubChem CID 78344
IUPAC Name 5-bromopyrimidine
SMILES C1=C(C=NC=N1)Br

Specifications

Melting Point 69°C to 74°C
Quantity 100 g
Beilstein 107326
Solubility Information Soluble in chloroform.
Formula Weight 158.99
Percent Purity 98%
Chemical Name or Material 5-Bromopyrimidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 224-992-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only