missing translation for 'onlineSavingsMsg'
Learn More

5-Bromopyrimidine-2-carboxylic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6404106

 View more versions of this product

Catalog No. AAH6404106


Only null left
Explore available promotions

Description

Description

5-Bromopyrimidine-2-carboxylic acid is employed as a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromopyrimidine-2-carboxylic acid is employed as a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Specifications

5-Bromopyrimidine-2-carboxylic acid
193°C to 198°C
MFCD00496793
121432
Slightly soluble in water.
C1=C(C=NC(=N1)C(=O)O)Br
202.995
203
Solid
37131-87-6
C5H3BrN2O2
5 g
5-bromo-pyrimidine-2-carboxylic acid, 5-bromo-2-pyrimidinecarboxylic acid, 2-pyrimidinecarboxylic acid, 5-bromo, 5-bromopyrimidine-2-carboxylicacid, pubchem16922, acmc-1agpr, ksc222c2t, 5-bromopyrimidine-2-caboxylic acid, abbypharma ap-10-1676, 2-pyrimidinecarboxylicacid, 5-bromo
XGPTXUYKEDPXCO-UHFFFAOYSA-N
5-bromopyrimidine-2-carboxylic acid
16462970
98%

Chemical Identifiers

37131-87-6
202.995
XGPTXUYKEDPXCO-UHFFFAOYSA-N
16462970
C1=C(C=NC(=N1)C(=O)O)Br
C5H3BrN2O2
MFCD00496793
5-bromo-pyrimidine-2-carboxylic acid, 5-bromo-2-pyrimidinecarboxylic acid, 2-pyrimidinecarboxylic acid, 5-bromo, 5-bromopyrimidine-2-carboxylicacid, pubchem16922, acmc-1agpr, ksc222c2t, 5-bromopyrimidine-2-caboxylic acid, abbypharma ap-10-1676, 2-pyrimidinecarboxylicacid, 5-bromo
5-bromopyrimidine-2-carboxylic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only