Learn More
5-Bromopyridine-3-sulfonyl chloride, 97%, Thermo Scientific™
$165.25 - $165.25
Chemical Identifiers
| CAS | 65001-21-0 |
|---|---|
| Molecular Formula | C5H3BrClNO2S |
| Molecular Weight (g/mol) | 256.498 |
| MDL Number | MFCD08235263 |
| InChI Key | AVILRJQPDYPXFQ-UHFFFAOYSA-N |
| Synonym | 5-bromopyridine-3-sulphonyl chloride, 5-bromo-3-pyridinesulfonyl chloride, 5-bromopyridine-3-sulfonylchloride, 5-bromo-pyridine-3-sulfonyl chloride, 5-bromo 3-pyridyl chlorosulfone, 5-bromopyridin-3-sulfonyl chloride, 3-bromopyridine-5-sulfonyl chloride, 5-bromopyridine-3-sulphonylchloride, 5-bromo-3-chlorosulphonyl pyridine, 3-pyridinesulfonylchloride, 5-bromo |
| PubChem CID | 20324474 |
| IUPAC Name | 5-bromopyridine-3-sulfonyl chloride |
| SMILES | C1=C(C=NC=C1Br)S(=O)(=O)Cl |
Chemical Identifiers
| 65001-21-0 | |
| 256.498 | |
| AVILRJQPDYPXFQ-UHFFFAOYSA-N | |
| 20324474 | |
| C1=C(C=NC=C1Br)S(=O)(=O)Cl |
| C5H3BrClNO2S | |
| MFCD08235263 | |
| 5-bromopyridine-3-sulphonyl chloride, 5-bromo-3-pyridinesulfonyl chloride, 5-bromopyridine-3-sulfonylchloride, 5-bromo-pyridine-3-sulfonyl chloride, 5-bromo 3-pyridyl chlorosulfone, 5-bromopyridin-3-sulfonyl chloride, 3-bromopyridine-5-sulfonyl chloride, 5-bromopyridine-3-sulphonylchloride, 5-bromo-3-chlorosulphonyl pyridine, 3-pyridinesulfonylchloride, 5-bromo | |
| 5-bromopyridine-3-sulfonyl chloride |
Specifications
| 65001-21-0 | |
| 100.0 | |
| Amber Glass Bottle | |
| MFCD08235263 | |
| 5-bromopyridine-3-sulphonyl chloride, 5-bromo-3-pyridinesulfonyl chloride, 5-bromopyridine-3-sulfonylchloride, 5-bromo-pyridine-3-sulfonyl chloride, 5-bromo 3-pyridyl chlorosulfone, 5-bromopyridin-3-sulfonyl chloride, 3-bromopyridine-5-sulfonyl chloride, 5-bromopyridine-3-sulphonylchloride, 5-bromo-3-chlorosulphonyl pyridine, 3-pyridinesulfonylchloride, 5-bromo | |
| C1=C(C=NC=C1Br)S(=O)(=O)Cl | |
| 256.498 | |
| 256.51 | |
| 5-Bromopyridine-3-sulfonyl chloride |
| 96.5 | |
| 97% | |
| C5H3BrClNO2S | |
| 250 mg | |
| AVILRJQPDYPXFQ-UHFFFAOYSA-N | |
| 5-bromopyridine-3-sulfonyl chloride | |
| 20324474 | |
| 97% |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger