missing translation for 'onlineSavingsMsg'
Learn More

5-Bromopyridine-3-sulfonyl chloride, 97%, Thermo Scientific™

Catalog No. CC67103CB
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC67103CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC67103CB Supplier Thermo Scientific Chemicals Supplier No. CC67103CB

Chemical Identifiers

CAS 65001-21-0
Molecular Formula C5H3BrClNO2S
Molecular Weight (g/mol) 256.498
MDL Number MFCD08235263
InChI Key AVILRJQPDYPXFQ-UHFFFAOYSA-N
Synonym 5-bromopyridine-3-sulphonyl chloride, 5-bromo-3-pyridinesulfonyl chloride, 5-bromopyridine-3-sulfonylchloride, 5-bromo-pyridine-3-sulfonyl chloride, 5-bromo 3-pyridyl chlorosulfone, 5-bromopyridin-3-sulfonyl chloride, 3-bromopyridine-5-sulfonyl chloride, 5-bromopyridine-3-sulphonylchloride, 5-bromo-3-chlorosulphonyl pyridine, 3-pyridinesulfonylchloride, 5-bromo
PubChem CID 20324474
IUPAC Name 5-bromopyridine-3-sulfonyl chloride
SMILES C1=C(C=NC=C1Br)S(=O)(=O)Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 256.51
Percent Purity 97%
Chemical Name or Material 5-Bromopyridine-3-sulfonyl chloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.