missing translation for 'onlineSavingsMsg'
Learn More

5-Bromopyridine-3-carboxaldehyde, 97%

CAS: 113118-81-3 | C6H4BrNO | 186.008 g/mol

Supplier:  Thermo Scientific Chemicals H5922406

 View more versions of this product

Catalog No. AAH5922406


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

5-Bromopyridine-3-carboxaldehyde
98°C to 102°C
MFCD03265758
Air Sensitive
NGUVGKAEOFPLDT-UHFFFAOYSA-N
5-bromopyridine-3-carbaldehyde
2784734
97%
113118-81-3
C6H4BrNO
5 g
5-bromonicotinaldehyde, 5-bromo-3-pyridinecarboxaldehyde, 5-bromo-3-formylpyridine, 3-bromopyridine-5-carboxaldehyde, 5-bromopyridine-3-carboxaldehyde, 5-bromo-pyridine-3-carbaldehyde, 3-bromo-5-formylpyridine, 3-pyridinecarboxaldehyde, 5-bromo, 3-bromopyridine-5-carbaldehyde, 5-bromo-3-pyridinecarbaldehyde
C1=C(C=NC=C1Br)C=O
186.008
186.01

Chemical Identifiers

113118-81-3
186.008
NGUVGKAEOFPLDT-UHFFFAOYSA-N
2784734
C1=C(C=NC=C1Br)C=O
C6H4BrNO
MFCD03265758
5-bromonicotinaldehyde, 5-bromo-3-pyridinecarboxaldehyde, 5-bromo-3-formylpyridine, 3-bromopyridine-5-carboxaldehyde, 5-bromopyridine-3-carboxaldehyde, 5-bromo-pyridine-3-carbaldehyde, 3-bromo-5-formylpyridine, 3-pyridinecarboxaldehyde, 5-bromo, 3-bromopyridine-5-carbaldehyde, 5-bromo-3-pyridinecarbaldehyde
5-bromopyridine-3-carbaldehyde
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H317
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c

H315-H317-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only