missing translation for 'onlineSavingsMsg'
Learn More

5-Bromopyridine-3-carboxaldehyde, 97%

Catalog No. AAH5922403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5922403 1 g
AAH5922406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5922403 Supplier Thermo Scientific Chemicals Supplier No. H5922403

CAS: 113118-81-3 | C6H4BrNO | 186.008 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 113118-81-3
Molecular Formula C6H4BrNO
Molecular Weight (g/mol) 186.008
MDL Number MFCD03265758
InChI Key NGUVGKAEOFPLDT-UHFFFAOYSA-N
Synonym 5-bromonicotinaldehyde, 5-bromo-3-pyridinecarboxaldehyde, 5-bromo-3-formylpyridine, 3-bromopyridine-5-carboxaldehyde, 5-bromopyridine-3-carboxaldehyde, 5-bromo-pyridine-3-carbaldehyde, 3-bromo-5-formylpyridine, 3-pyridinecarboxaldehyde, 5-bromo, 3-bromopyridine-5-carbaldehyde, 5-bromo-3-pyridinecarbaldehyde
PubChem CID 2784734
IUPAC Name 5-bromopyridine-3-carbaldehyde
SMILES C1=C(C=NC=C1Br)C=O

Specifications

Melting Point 98°C to 102°C
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 186.01
Percent Purity 97%
Chemical Name or Material 5-Bromopyridine-3-carboxaldehyde
Hazard Category H315-H317-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H317
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only