missing translation for 'onlineSavingsMsg'
Learn More

5-Bromopyridine-2-carboxaldehyde, 99%

Catalog No. AAH5015403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5015403 1 g
AAH5015406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5015403 Supplier Thermo Scientific Chemicals Supplier No. H5015403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 31181-90-5 | C6H4BrNO | 186.008 g/mol

5-Bromopyridine-2-carboxaldehyde is used as organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromopyridine-2-carboxaldehyde is used as organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Air sensitive. Store away from oxidizing agents.

Chemical Identifiers

CAS 31181-90-5
Molecular Formula C6H4BrNO
Molecular Weight (g/mol) 186.008
MDL Number MFCD04112535
InChI Key ZQVLPMNLLKGGIU-UHFFFAOYSA-N
Synonym 5-bromopicolinaldehyde, 5-bromo-2-formylpyridine, 5-bromo-2-pyridinecarboxaldehyde, 5-bromopyridine-2-carboxaldehyde, 5-bromo-pyridine-2-carbaldehyde, 5-bromo-2-pyridinecarbaldehyde, 2-pyridinecarboxaldehyde, 5-bromo, 5-bromo-2-pyridine-carbaldehyde, pubchem5059, 5-bromo picolinaldehyde
PubChem CID 9877562
IUPAC Name 5-bromopyridine-2-carbaldehyde
SMILES C1=CC(=NC=C1Br)C=O

Specifications

Melting Point 94°C to 96°C
Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 186.01
Percent Purity 99%
Chemical Name or Material 5-Bromopyridine-2-carboxaldehyde
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only