missing translation for 'onlineSavingsMsg'
Learn More

5-Bromopyrazine-2-carboxylic acid, 95%

Catalog No. p-7057056
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
250 mg
Catalog No. Quantity
AAH66382MD 250 mg
AAH6638203 1 g
2 options

Catalog No. AAH66382MD

Supplier: Thermo Scientific Chemicals H66382MD

Only null left

CAS: 876161-05-6 | C5H3BrN2O2 | 203.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 876161-05-6
Molecular Formula C5H3BrN2O2
Molecular Weight (g/mol) 203.00
MDL Number MFCD09909672
InChI Key NPJHCKULJAIWGV-UHFFFAOYSA-N
PubChem CID 18779790
IUPAC Name 5-bromopyrazine-2-carboxylic acid
SMILES OC(=O)C1=CN=C(Br)C=N1

Specifications

Quantity 250 mg
Synonym 5-bromo-2-pyrazinecarboxylic acid, 2-pyrazinecarboxylicacid, 5-bromo, 2-pyrazinecarboxylic acid, 5-bromo, 5-bromo-2-pyrazinecarboxylicacid, ksc446e0j, 2-pyrazinecarboxylicacid,5-bromo, 2-bromopyrazine-5-carboxylic acid
Formula Weight 203
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 5-Bromopyrazine-2-carboxylic acid
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only