missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromooxindole 97.0+%, TCI America™
$74.56 - $331.81
Chemical Identifiers
| CAS | 20870-78-4 |
|---|---|
| Molecular Formula | C8H6BrNO |
| Molecular Weight (g/mol) | 212.046 |
| MDL Number | MFCD00456998 |
| InChI Key | VIMNAEVMZXIKFL-UHFFFAOYSA-N |
| Synonym | 5-bromooxindole, 5-bromoindolin-2-one, 5-bromo-2-oxindole, 5-bromo-1,3-dihydro-2h-indol-2-one, 5-bromo-2,3-dihydro-1h-indol-2-one, 5-bromo-2-indolinone, 5-bromo-1,3-dihydro-indol-2-one, 2h-indol-2-one, 5-bromo-1,3-dihydro, 5-bromo-2-oxindol, 1,3-dihydro-5-bromoindol-2-one |
| PubChem CID | 611193 |
| IUPAC Name | 5-bromo-1,3-dihydroindol-2-one |
| SMILES | C1C2=C(C=CC(=C2)Br)NC1=O |
Chemical Identifiers
| 20870-78-4 | |
| 212.046 | |
| VIMNAEVMZXIKFL-UHFFFAOYSA-N | |
| 611193 | |
| C1C2=C(C=CC(=C2)Br)NC1=O |
| C8H6BrNO | |
| MFCD00456998 | |
| 5-bromooxindole, 5-bromoindolin-2-one, 5-bromo-2-oxindole, 5-bromo-1,3-dihydro-2h-indol-2-one, 5-bromo-2,3-dihydro-1h-indol-2-one, 5-bromo-2-indolinone, 5-bromo-1,3-dihydro-indol-2-one, 2h-indol-2-one, 5-bromo-1,3-dihydro, 5-bromo-2-oxindol, 1,3-dihydro-5-bromoindol-2-one | |
| 5-bromo-1,3-dihydroindol-2-one |
Specifications
| 20870-78-4 | |
| C8H6BrNO | |
| 1 g | |
| VIMNAEVMZXIKFL-UHFFFAOYSA-N | |
| 5-bromo-1,3-dihydroindol-2-one | |
| 611193 | |
| ≥97.0% (GC,N) | |
| 5-Bromooxindole |
| 222°C | |
| MFCD00456998 | |
| 5-bromooxindole, 5-bromoindolin-2-one, 5-bromo-2-oxindole, 5-bromo-1,3-dihydro-2h-indol-2-one, 5-bromo-2,3-dihydro-1h-indol-2-one, 5-bromo-2-indolinone, 5-bromo-1,3-dihydro-indol-2-one, 2h-indol-2-one, 5-bromo-1,3-dihydro, 5-bromo-2-oxindol, 1,3-dihydro-5-bromoindol-2-one | |
| C1C2=C(C=CC(=C2)Br)NC1=O | |
| 212.046 | |
| 212.05 | |
| Crystalline Powder |
Safety and Handling
TSCA : No