missing translation for 'onlineSavingsMsg'
Learn More

5-Bromooxindole 97.0+%, TCI America™

Catalog No. B37411G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37411G 1 g
B37415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B37411G Supplier TCI America Supplier No. B37411G

Chemical Identifiers

CAS 20870-78-4
Molecular Formula C8H6BrNO
Molecular Weight (g/mol) 212.046
MDL Number MFCD00456998
InChI Key VIMNAEVMZXIKFL-UHFFFAOYSA-N
Synonym 5-bromooxindole, 5-bromoindolin-2-one, 5-bromo-2-oxindole, 5-bromo-1,3-dihydro-2h-indol-2-one, 5-bromo-2,3-dihydro-1h-indol-2-one, 5-bromo-2-indolinone, 5-bromo-1,3-dihydro-indol-2-one, 2h-indol-2-one, 5-bromo-1,3-dihydro, 5-bromo-2-oxindol, 1,3-dihydro-5-bromoindol-2-one
PubChem CID 611193
IUPAC Name 5-bromo-1,3-dihydroindol-2-one
SMILES C1C2=C(C=CC(=C2)Br)NC1=O

Specifications

Melting Point 222°C
Quantity 1 g
Formula Weight 212.05
Percent Purity ≥97.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromooxindole