missing translation for 'onlineSavingsMsg'
Learn More

5-Bromooxindole, 98%

Catalog No. AAH5057303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5057303 1 g
AAH5057306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5057303 Supplier Thermo Scientific Chemicals Supplier No. H5057303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 20870-78-4 | C8H6BrNO | 212.046 g/mol

5-Bromooxindole is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromooxindole is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in Dimethylformamide.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.

Chemical Identifiers

CAS 20870-78-4
Molecular Formula C8H6BrNO
Molecular Weight (g/mol) 212.046
MDL Number MFCD00456998
InChI Key VIMNAEVMZXIKFL-UHFFFAOYSA-N
Synonym 5-bromooxindole, 5-bromoindolin-2-one, 5-bromo-2-oxindole, 5-bromo-1,3-dihydro-2h-indol-2-one, 5-bromo-2,3-dihydro-1h-indol-2-one, 5-bromo-2-indolinone, 5-bromo-1,3-dihydro-indol-2-one, 2h-indol-2-one, 5-bromo-1,3-dihydro, 5-bromo-2-oxindol, 1,3-dihydro-5-bromoindol-2-one
PubChem CID 611193
IUPAC Name 5-bromo-1,3-dihydroindol-2-one
SMILES C1C2=C(C=CC(=C2)Br)NC1=O

Specifications

Quantity 1 g
Solubility Information Soluble in Dimethylformamide.
Formula Weight 212.05
Percent Purity 98%
Chemical Name or Material 5-Bromooxindole
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only