missing translation for 'onlineSavingsMsg'
Learn More

5-Bromooxindole, 97%, Thermo Scientific™

Chemical Identifiers

CAS 20870-78-4
Molecular Formula C8H6BrNO
Molecular Weight (g/mol) 212.046
InChI Key VIMNAEVMZXIKFL-UHFFFAOYSA-N
Synonym 5-bromooxindole, 5-bromoindolin-2-one, 5-bromo-2-oxindole, 5-bromo-1,3-dihydro-2h-indol-2-one, 5-bromo-2,3-dihydro-1h-indol-2-one, 5-bromo-2-indolinone, 5-bromo-1,3-dihydro-indol-2-one, 2h-indol-2-one, 5-bromo-1,3-dihydro, 5-bromo-2-oxindol, 1,3-dihydro-5-bromoindol-2-one
PubChem CID 611193
IUPAC Name 5-bromo-1,3-dihydroindol-2-one
SMILES C1C2=C(C=CC(=C2)Br)NC1=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
S05248DA
View Documents
N/A 1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

20870-78-4
212.046
5-bromooxindole, 5-bromoindolin-2-one, 5-bromo-2-oxindole, 5-bromo-1,3-dihydro-2h-indol-2-one, 5-bromo-2,3-dihydro-1h-indol-2-one, 5-bromo-2-indolinone, 5-bromo-1,3-dihydro-indol-2-one, 2h-indol-2-one, 5-bromo-1,3-dihydro, 5-bromo-2-oxindol, 1,3-dihydro-5-bromoindol-2-one
5-bromo-1,3-dihydroindol-2-one
C8H6BrNO
VIMNAEVMZXIKFL-UHFFFAOYSA-N
611193
C1C2=C(C=CC(=C2)Br)NC1=O

Specifications

20870-78-4
C8H6BrNO
VIMNAEVMZXIKFL-UHFFFAOYSA-N
5-bromo-1,3-dihydroindol-2-one
611193
97%
97%
5-bromooxindole, 5-bromoindolin-2-one, 5-bromo-2-oxindole, 5-bromo-1,3-dihydro-2h-indol-2-one, 5-bromo-2,3-dihydro-1h-indol-2-one, 5-bromo-2-indolinone, 5-bromo-1,3-dihydro-indol-2-one, 2h-indol-2-one, 5-bromo-1,3-dihydro, 5-bromo-2-oxindol, 1,3-dihydro-5-bromoindol-2-one
C1C2=C(C=CC(=C2)Br)NC1=O
212.046
212.05
5-Bromo-1, 3-dihydro-2H-indol-2-one
Videos
Safety and Handling

Safety and Handling

Warning

SDS
Documents

Documents