missing translation for 'onlineSavingsMsg'
Learn More

5-Bromonicotinamide 98.0+%, TCI America™

Catalog No. B35365G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B35365G Supplier TCI America Supplier No. B35365G
Only null left

Chemical Identifiers

CAS 28733-43-9
Molecular Formula C6H5BrN2O
Molecular Weight (g/mol) 201.023
MDL Number MFCD00173919
InChI Key YOQRXZIMSKLRCY-UHFFFAOYSA-N
Synonym 5-bromonicotinamide, 5-bromo-3-pyridinecarboxamide, nicotinamide, 5-bromo, 3-pyridinecarboxamide, 5-bromo, 5-bromo-nicotinamide, brt, pubchem1212, acmc-1cfs0, 3-bromo-5-carbamoylpyridine, 5-22-02-00182 beilstein handbook reference
PubChem CID 1808
IUPAC Name 5-bromopyridine-3-carboxamide
SMILES C1=C(C=NC=C1Br)C(=O)N

Specifications

Melting Point 219°C
Color White-Yellow
Quantity 5 g
Formula Weight 201.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromonicotinamide
RTECSNumber QS4170000
TSCA No