missing translation for 'onlineSavingsMsg'
Learn More

5-(Bromothythyl)-3-phénylisoxazole, 97%, Thermo Scientific™

Numéro de catalogue. CC64508DA
Modifier l'affichage
Click to view available options
Quantité:
1 g
Emballage:
Bouteille en verre ambrée
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
CC64508DA 1 g Bouteille en verre ambrée
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. CC64508DA Fournisseur Thermo Scientific Chemicals Code fournisseur CC64508DA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 2039-50-1
Formule moléculaire C10H8BrNO
Poids moléculaire (g/mol) 238.084
Numéro MDL MFCD00159724
Clé InChI ANRMBFFXQKJEIS-UHFFFAOYSA-N
PubChem CID 2748796
Nom de l’IUPAC 5-(bromométhyl)-3-phényl-1,2-oxazole
SOURIRES C1=CC=C(C=C1)C2=NOC(=C2)CBr

Spécifications

Nom ou substance chimique 5-(Bromomethyl)-3-phenylisoxazole
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Bouteille en verre ambrée
Quantité 1 g
Synonyme 5-bromomethyl-3-phenylisoxazole, 5-bromomethyl-3-phenyl-1,2-oxazole, isoxazole, 5-bromomethyl-3-phenyl, 5-bromomethyl-3-phenyl-isoxazole, 3-phenyl-5-bromomethylisoxazole, isoxazole,5-bromomethyl-3-phenyl
Poids de formule 238.08
Pourcentage de pureté 97%
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.