missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-(Bromomethyl)-1-methyl-1H-1,2,3-benzotriazole, 90+%, Thermo Scientific™
Specifications
Specifications
| Assay Percent Range | 95% |
| Packaging | Amber Glass Bottle |
| Quantity | 1 g |
| Formula Weight | 226.08 |
| Percent Purity | 95% |
| Chemical Name or Material | 5-(Bromomethyl)-1-methyl-1H-1, 2, 3-benzotriazole |