missing translation for 'onlineSavingsMsg'
Learn More

5-(Bromomethyl)-1-methyl-1H-1,2,3-benzotriazole, 90+%, Thermo Scientific™

Catalog No. CC23508CB
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC23508CB 250 mg Amber Glass Bottle
CC23508DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC23508CB Supplier Thermo Scientific Chemicals Supplier No. CC23508CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 499770-76-2
Molecular Formula C8H8BrN3
Molecular Weight (g/mol) 226.077
InChI Key OSUZHHPMRAIJDY-UHFFFAOYSA-N
Synonym 5-bromomethyl-1-methyl-1h-benzo d 1,2,3 triazole, 5-bromomethyl-1-methyl-1h-1,2,3-benzotriazole, 5-bromomethyl-1-methylbenzotriazole, 5-bromomethyl-1-methyl-benzotriazole, 1h-benzotriazole, 5-bromomethyl-1-methyl, 5-bromomethyl-1-methyl-1,2,3-benzotriazole, 5-bromomethyl-1-methyl-1h-benzotriazole
PubChem CID 2795418
IUPAC Name 5-(bromomethyl)-1-methylbenzotriazole
SMILES CN1C2=C(C=C(C=C2)CBr)N=N1

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 226.08
Percent Purity 95%
Chemical Name or Material 5-(Bromomethyl)-1-methyl-1H-1, 2, 3-benzotriazole