missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromoisoquinoline 96.0+%, TCI America™
$63.38 - $186.41
Chemical Identifiers
| CAS | 34784-04-8 |
|---|---|
| Molecular Formula | C9H6BrN |
| Molecular Weight (g/mol) | 208.06 |
| MDL Number | MFCD01646405 |
| InChI Key | CYJZJGYYTFQQBY-UHFFFAOYSA-N |
| Synonym | isoquinoline, 5-bromo, 5-bromo-isoquinoline, 5-bromo isoquinoline, 5-bromoisoquinoline?, pubchem6214, acmc-1adiy, 5-bromoisoquinoline, ksc226k2l, buttpark 44\07-76, 5-bromoisoquinoline 250mg |
| PubChem CID | 736487 |
| IUPAC Name | 5-bromoisoquinoline |
| SMILES | BrC1=C2C=CN=CC2=CC=C1 |
Chemical Identifiers
| 34784-04-8 | |
| 208.06 | |
| CYJZJGYYTFQQBY-UHFFFAOYSA-N | |
| 736487 | |
| BrC1=C2C=CN=CC2=CC=C1 |
| C9H6BrN | |
| MFCD01646405 | |
| isoquinoline, 5-bromo, 5-bromo-isoquinoline, 5-bromo isoquinoline, 5-bromoisoquinoline?, pubchem6214, acmc-1adiy, 5-bromoisoquinoline, ksc226k2l, buttpark 44\07-76, 5-bromoisoquinoline 250mg | |
| 5-bromoisoquinoline |
Specifications
| 34784-04-8 | |
| Yellow | |
| C9H6BrN | |
| 1 g | |
| CYJZJGYYTFQQBY-UHFFFAOYSA-N | |
| 5-bromoisoquinoline | |
| 736487 | |
| ≥96.0% (GC,T) | |
| 5-Bromoisoquinoline |
| 82°C | |
| 97°C | |
| MFCD01646405 | |
| isoquinoline, 5-bromo, 5-bromo-isoquinoline, 5-bromo isoquinoline, 5-bromoisoquinoline?, pubchem6214, acmc-1adiy, 5-bromoisoquinoline, ksc226k2l, buttpark 44\07-76, 5-bromoisoquinoline 250mg | |
| BrC1=C2C=CN=CC2=CC=C1 | |
| 208.06 | |
| 208.06 | |
| Crystalline Powder |
Safety and Handling
TSCA : No