missing translation for 'onlineSavingsMsg'
Learn More

5-Bromoisophthalic Acid 98.0+%, TCI America™

Catalog No. B42321G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B42321G 1 g
B42325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B42321G Supplier TCI America Supplier No. B42321G
Only null left

Chemical Identifiers

CAS 23351-91-9
Molecular Formula C8H3BrO4
Molecular Weight (g/mol) 243.01
MDL Number MFCD00152019
InChI Key JATKASGNRMGFSW-UHFFFAOYSA-L
Synonym 5-bromoisophthalic acid, 1,3-benzenedicarboxylic acid, 5-bromo, 5-bromoisophthalicacid, pubchem22068, 5-bromo-isophthalic acid, acmc-209g2n, ksc202c0n, 5-bromo-1,3-benzenedicarboxylic acid, 1,3-benzenedicarboxylicacid, 5-bromo
PubChem CID 90073
IUPAC Name 5-bromobenzene-1,3-dicarboxylate
SMILES [O-]C(=O)C1=CC(=CC(Br)=C1)C([O-])=O

Specifications

Melting Point 287°C
Color White
Quantity 1 g
Formula Weight 245.03
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromoisophthalic Acid
EINECSNumber (3)-0062
TSCA Yes