missing translation for 'onlineSavingsMsg'
Learn More

5-Bromoindoxyl diacetate, 98+%, Thermo Scientific Chemicals

Catalog No. AAL1118903
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1118903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1118903 Supplier Thermo Scientific Chemicals Supplier No. L1118903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 33588-54-4
Molecular Formula C12H10BrNO3
Molecular Weight (g/mol) 296.12
MDL Number MFCD00005799
InChI Key XJRIDJAGAYGJCK-UHFFFAOYSA-N
Synonym 5-bromoindoxyl diacetate, 1-acetyl-5-bromo-1h-indol-3-yl acetate, 5-bromoindoxyldiacetate, n-acetyl-5-bromoindolyl acetate, 1-acetyl-3-acetyloxy-5-bromoindole, 3-acetoxy-1-acetyl-5-bromoindole, 1h-indol-3-ol, 1-acetyl-5-bromo-, acetate ester, 3-acetoxy-1-acetyl-5-bromo-indole, 1h-indol-3-ol, 1-acetyl-5-bromo-, acetate, pubchem16263
PubChem CID 118505
IUPAC Name (1-acetyl-5-bromoindol-3-yl) acetate
SMILES CC(=O)N1C=C(C2=C1C=CC(=C2)Br)OC(=O)C

Specifications

Melting Point 122°C to 125°C
Quantity 1 g
Beilstein 236935
Formula Weight 296.13
Percent Purity ≥98%
Chemical Name or Material 5-Bromoindoxyl diacetate
EINECSNumber 251-584-0
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only