missing translation for 'onlineSavingsMsg'
Learn More

5-Bromoindoline 98.0+%, TCI America™

Catalog No. B38851G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38851G 1 g
B38855G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B38851G Supplier TCI America Supplier No. B38851G

Chemical Identifiers

CAS 22190-33-6
Molecular Formula C8H8BrN
Molecular Weight (g/mol) 198.063
MDL Number MFCD00027410
InChI Key QEDCHCLHHGGYBT-UHFFFAOYSA-N
Synonym 5-bromoindoline, 1h-indole, 5-bromo-2,3-dihydro, 5-bromo-indoline, zlchem 28, 5-bromodihydroindole, pubchem7319, acmc-1cez6, 5-bromo-2,3-dihydro-indole, ksc497i6r
PubChem CID 3411566
IUPAC Name 5-bromo-2,3-dihydro-1H-indole
SMILES C1CNC2=C1C=C(C=C2)Br

Specifications

Melting Point 41°C
Boiling Point 132°C
Quantity 1 g
Formula Weight 198.06
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromoindoline
TSCA No
Recommended Storage Freezer