missing translation for 'onlineSavingsMsg'
Learn More

5-Bromoindoline, 98+%

Catalog No. p-7058467
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1699203 1 g
AAL1699206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1699203 Supplier Thermo Scientific Chemicals Supplier No. L1699203

CAS: 22190-33-6 | C8H8BrN | 198.063 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22190-33-6
Molecular Formula C8H8BrN
Molecular Weight (g/mol) 198.063
MDL Number MFCD00027410
InChI Key QEDCHCLHHGGYBT-UHFFFAOYSA-N
Synonym 5-bromoindoline, 1h-indole, 5-bromo-2,3-dihydro, 5-bromo-indoline, zlchem 28, 5-bromodihydroindole, pubchem7319, acmc-1cez6, 5-bromo-2,3-dihydro-indole, ksc497i6r
PubChem CID 3411566
IUPAC Name 5-bromo-2,3-dihydro-1H-indole
SMILES C1CNC2=C1C=C(C=C2)Br

Specifications

Melting Point 36°C to 40°C
Boiling Point 114°C to 115°C (5 mmHg)
Quantity 1 g
Beilstein 121590
Formula Weight 198.06
Percent Purity ≥98%
Chemical Name or Material 5-Bromoindoline
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only