missing translation for 'onlineSavingsMsg'
Learn More

5-Bromoindole 99.0+%, TCI America™

Catalog No. p-7119233
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
B17385G 5 g
B173825G 25 g
2 options

Catalog No. B17385G

Supplier: TCI America B17385G

Only null left

Chemical Identifiers

CAS 10075-50-0
Molecular Formula C8H6BrN
Molecular Weight (g/mol) 196.05
MDL Number MFCD00005670
InChI Key VXWVFZFZYXOBTA-UHFFFAOYSA-N
Synonym 5-bromoindole, 1h-indole, 5-bromo, 5-bromo indole, 5-bi, 5bromoindole, 5-bromo-indole, zlchem 227, 5-bromo 1h indole, pubchem1661
PubChem CID 24905
IUPAC Name 5-bromo-1H-indole
SMILES BrC1=CC=C2NC=CC2=C1

Specifications

Melting Point 90°C
Quantity 5 g
Formula Weight 196.05
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromoindole