missing translation for 'onlineSavingsMsg'
Learn More

5-Bromoindole, 99%

Catalog No. AC107061000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC107061000 100 g Glass bottle
AC107060050 5 g Glass bottle
AC107060250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC107061000 Supplier Thermo Scientific Chemicals Supplier No. 107061000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10075-50-0 | C8H6BrN | 196.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10075-50-0
Molecular Formula C8H6BrN
Molecular Weight (g/mol) 196.05
MDL Number MFCD00005670
InChI Key VXWVFZFZYXOBTA-UHFFFAOYSA-N
Synonym 5-bromoindole, 1h-indole, 5-bromo, 5-bromo indole, 5-bi, 5bromoindole, 5-bromo-indole, zlchem 227, 5-bromo 1h indole, pubchem1661
PubChem CID 24905
IUPAC Name 5-bromo-1H-indole
SMILES BrC1=CC=C2NC=CC2=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 89.0°C to 92.0°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 100 g
Formula Weight 196.05
Percent Purity 99%
Physical Form Chunks and/or Powder
Chemical Name or Material 5-Bromoindole, 99%
Show More Show Less
EINECSNumber 233-208-7

RUO – Research Use Only