missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromoindole-3-ethanol 98.0+%, TCI America™
$123.03 - $369.08
Chemical Identifiers
| CAS | 32774-29-1 |
|---|---|
| Molecular Formula | C10H10BrNO |
| Molecular Weight (g/mol) | 240.1 |
| MDL Number | MFCD00130167 |
| InChI Key | ZENXDUDCTZLSRP-UHFFFAOYSA-N |
| Synonym | 5-Bromo-3-(2-hydroxyethyl)indole |
| PubChem CID | 122956 |
| IUPAC Name | 2-(5-bromo-1H-indol-3-yl)ethanol |
| SMILES | C1=CC2=C(C=C1Br)C(=CN2)CCO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B5068200MG
|
TCI America
B5068200MG |
200 mg |
Each for $123.03
|
|
|||||
|
B50681G
|
TCI America
B50681G |
1 g |
Each for $369.08
|
|
|||||
Chemical Identifiers
| 32774-29-1 | |
| 240.1 | |
| ZENXDUDCTZLSRP-UHFFFAOYSA-N | |
| 122956 | |
| C1=CC2=C(C=C1Br)C(=CN2)CCO |
| C10H10BrNO | |
| MFCD00130167 | |
| 5-Bromo-3-(2-hydroxyethyl)indole | |
| 2-(5-bromo-1H-indol-3-yl)ethanol |
Specifications
| 32774-29-1 | |
| White-Yellow | |
| MFCD00130167 | |
| 5-Bromo-3-(2-hydroxyethyl)indole | |
| C1=CC2=C(C=C1Br)C(=CN2)CCO | |
| 240.1 | |
| 240.10 | |
| Crystalline Powder |
| 85°C | |
| C10H10BrNO | |
| 200 mg | |
| ZENXDUDCTZLSRP-UHFFFAOYSA-N | |
| 2-(5-bromo-1H-indol-3-yl)ethanol | |
| 122956 | |
| ≥98.0% (GC) | |
| 5-Bromoindole-3-ethanol |
Safety and Handling
TSCA : No