missing translation for 'onlineSavingsMsg'
Learn More

5-Bromoindole-3-carboxaldehyde 98.0+%, TCI America™

Catalog No. p-7123784
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48401G 1 g
B48405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B48401G Supplier TCI America Supplier No. B48401G

Chemical Identifiers

CAS 877-03-2
Molecular Formula C9H6BrNO
Molecular Weight (g/mol) 224.057
MDL Number MFCD00152016
InChI Key PEENKJZANBYXNB-UHFFFAOYSA-N
Synonym 5-bromoindole-3-carboxaldehyde, 5-bromoindole-3-carbaldehyde, 5-bromoindole-3-carboxyaldehyde, 5-bromo-3-formylindole, 5-bromoindole-3-aldehyde, 5-bromo-3-formyl-1h-indole, 5-bromo-1h-indole-3-carboxaldehyde, 1h-indole-3-carboxaldehyde, 5-bromo, pubchem9122, acmc-209qpj
PubChem CID 70137
IUPAC Name 5-bromo-1H-indole-3-carbaldehyde
SMILES C1=CC2=C(C=C1Br)C(=CN2)C=O

Specifications

Melting Point 206°C
Color White-Yellow
Quantity 1 g
Formula Weight 224.06
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromoindole-3-carboxaldehyde