missing translation for 'onlineSavingsMsg'
Learn More

5-Bromoindole-2-carboxylic Acid 98.0+%, TCI America™

Catalog No. B38361G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38361G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B38361G Supplier TCI America Supplier No. B38361G

Chemical Identifiers

CAS 7254-19-5
Molecular Formula C9H6BrNO2
Molecular Weight (g/mol) 240.056
MDL Number MFCD00022705
InChI Key YAULOOYNCJDPPU-UHFFFAOYSA-N
Synonym 5-bromoindole-2-carboxylic acid, 5-bromo-2-indolecarboxylic acid, 1h-indolecarboxylic acid, 5-bromo, 1h-indole-2-carboxylic acid, 5-bromo, pubchem1663, zlchem 1343, acmc-1bb5x, chembl22435, 5-bromo indole carboxylic acid, 5-bromanyl-1h-indole-2-carboxylic acid
PubChem CID 252137
IUPAC Name 5-bromo-1H-indole-2-carboxylic acid
SMILES C1=CC2=C(C=C1Br)C=C(N2)C(=O)O

Specifications

Melting Point 280°C
Quantity 1 g
Formula Weight 240.06
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromoindole-2-carboxylic Acid
TSCA No
Recommended Storage Freezer