Learn More
5-Bromoindole-2-carboxylic acid, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 398652500
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| 5-Bromoindole-2-carboxylic acid | |
| 97.5 | |
| 97.5% min. (HPLC) | |
| C9H6BrNO2 | |
| 250mg | |
| YAULOOYNCJDPPU-UHFFFAOYSA-N | |
| 5-bromo-1H-indole-2-carboxylic acid | |
| 252137 | |
| 98% |
| 7254-19-5 | |
| 100.0 | |
| Glass Bottle | |
| MFCD00022705 | |
| 5-bromoindole-2-carboxylic acid, 5-bromo-2-indolecarboxylic acid, 1h-indolecarboxylic acid, 5-bromo, 1h-indole-2-carboxylic acid, 5-bromo, pubchem1663, zlchem 1343, acmc-1bb5x, chembl22435, 5-bromo indole carboxylic acid, 5-bromanyl-1h-indole-2-carboxylic acid | |
| C1=CC2=C(C=C1Br)C=C(N2)C(=O)O | |
| 240.05 | |
| 240.05 |
Chemical Identifiers
| 7254-19-5 | |
| 240.05 | |
| YAULOOYNCJDPPU-UHFFFAOYSA-N | |
| 252137 | |
| C1=CC2=C(C=C1Br)C=C(N2)C(=O)O |
| C9H6BrNO2 | |
| MFCD00022705 | |
| 5-bromoindole-2-carboxylic acid, 5-bromo-2-indolecarboxylic acid, 1h-indolecarboxylic acid, 5-bromo, 1h-indole-2-carboxylic acid, 5-bromo, pubchem1663, zlchem 1343, acmc-1bb5x, chembl22435, 5-bromo indole carboxylic acid, 5-bromanyl-1h-indole-2-carboxylic acid | |
| 5-bromo-1H-indole-2-carboxylic acid |
Safety and Handling
GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO â Research Use Only