missing translation for 'onlineSavingsMsg'
Learn More

5-Bromoindole-2-carboxylic acid, 98%

Catalog No. AC398650010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC398650010 1 g Glass bottle
AC398650050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC398650010 Supplier Thermo Scientific Chemicals Supplier No. 398650010

CAS: 7254-19-5 | C9H6BrNO2 | 240.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7254-19-5
Molecular Formula C9H6BrNO2
Molecular Weight (g/mol) 240.05
MDL Number MFCD00022705
InChI Key YAULOOYNCJDPPU-UHFFFAOYSA-N
Synonym 5-bromoindole-2-carboxylic acid, 5-bromo-2-indolecarboxylic acid, 1h-indolecarboxylic acid, 5-bromo, 1h-indole-2-carboxylic acid, 5-bromo, pubchem1663, zlchem 1343, acmc-1bb5x, chembl22435, 5-bromo indole carboxylic acid, 5-bromanyl-1h-indole-2-carboxylic acid
PubChem CID 252137
IUPAC Name 5-bromo-1H-indole-2-carboxylic acid
SMILES C1=CC2=C(C=C1Br)C=C(N2)C(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 240.05
Percent Purity 98%
Chemical Name or Material 5-Bromoindole-2-carboxylic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only