missing translation for 'onlineSavingsMsg'
Learn More

5-Bromoindole-2-carboxylic acid, 96%

Catalog No. p-7057065
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6639603 1 g
AAH6639606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6639603 Supplier Thermo Scientific Chemicals Supplier No. H6639603
Only null left

CAS: 7254-19-5 | C9H6BrNO2 | 240.056 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7254-19-5
Molecular Formula C9H6BrNO2
Molecular Weight (g/mol) 240.056
MDL Number MFCD00022705
InChI Key YAULOOYNCJDPPU-UHFFFAOYSA-N
Synonym 5-bromoindole-2-carboxylic acid, 5-bromo-2-indolecarboxylic acid, 1h-indolecarboxylic acid, 5-bromo, 1h-indole-2-carboxylic acid, 5-bromo, pubchem1663, zlchem 1343, acmc-1bb5x, chembl22435, 5-bromo indole carboxylic acid, 5-bromanyl-1h-indole-2-carboxylic acid
PubChem CID 252137
IUPAC Name 5-bromo-1H-indole-2-carboxylic acid
SMILES C1=CC2=C(C=C1Br)C=C(N2)C(=O)O

Specifications

Quantity 1 g
Beilstein 153253
Formula Weight 240.06
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 5-Bromoindole-2-carboxylic acid
Hazard Category H302+H312-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only