missing translation for 'onlineSavingsMsg'
Learn More

5-Bromoindazole 97.0+%, TCI America™

Catalog No. B37851G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37851G 1 g
B37855G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B37851G Supplier TCI America Supplier No. B37851G

Chemical Identifiers

CAS 53857-57-1
Molecular Formula C7H5BrN2
Molecular Weight (g/mol) 197.04
MDL Number MFCD00839493,MFCD26227374
InChI Key STVHMYNPQCLUNJ-UHFFFAOYSA-N
Synonym 5-bromoindazole, 5-bromo-2h-indazole, 1h-indazole, 5-bromo, 5-bromo-1hindazole, chembl16425, 5-brom-1h-indazol, 5-bromoazaindole, 5-bromo indazole, 5-bromo-indazole
PubChem CID 761929
IUPAC Name 5-bromo-1H-indazole
SMILES BrC1=CC=C2NN=CC2=C1

Specifications

Melting Point 132°C
Quantity 1 g
Formula Weight 197.04
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromoindazole