missing translation for 'onlineSavingsMsg'
Learn More

5-Bromoimidazo[1,2-a]pyridine, 97%

Catalog No. AAH3324003
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3324003 1 g
AAH3324009 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3324003 Supplier Thermo Scientific Chemicals Supplier No. H3324003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 69214-09-1 | C7H5BrN2 | 197.035 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 69214-09-1
Molecular Formula C7H5BrN2
Molecular Weight (g/mol) 197.035
MDL Number MFCD08275698
InChI Key CCOFGVWHMYYDBG-UHFFFAOYSA-N
Synonym 5-bromoimidazo 1,2-a pyridine, 5-bromo-imidazo 1,2-a pyridine, 5-bromoh-imidazo 1,2-a pyridine, imidazo 1,2-a pyridine, 5-bromo, acmc-209o6n, 5-bromo-4-hydroimidazo 1,2-a pyridine
PubChem CID 13037241
IUPAC Name 5-bromoimidazo[1,2-a]pyridine
SMILES C1=CC2=NC=CN2C(=C1)Br

Specifications

Melting Point 61°C to 64°C
Quantity 1 g
Formula Weight 197.03
Percent Purity ≥97%
Chemical Name or Material 5-Bromoimidazo[1,2-a]pyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only