missing translation for 'onlineSavingsMsg'
Learn More

5-Bromobenzofuran 97.0+%, TCI America™

Catalog No. p-7123225
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
B38891G 1 g
B38895G 5 g
2 options

Catalog No. B38891G

Supplier: TCI America B38891G

Only null left

Chemical Identifiers

CAS 23145-07-5
Molecular Formula C8H5BrO
Molecular Weight (g/mol) 197.031
MDL Number MFCD03407317
InChI Key AYOVPQORFBWFNO-UHFFFAOYSA-N
Synonym 5-bromobenzofuran, benzofuran, 5-bromo, 5-bromobenzo b furan, 5-bromo-benzofuran, 5-bromobenzo beta furan, buttpark 95\50-66, pubchem22017, 5-bromobenzo b-furan, #
PubChem CID 90015
IUPAC Name 5-bromo-1-benzofuran
SMILES C1=CC2=C(C=CO2)C=C1Br

Specifications

Melting Point 8°C
Color Yellow
Boiling Point 226°C
Quantity 1 g
Formula Weight 197.03
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Bromobenzofuran