missing translation for 'onlineSavingsMsg'
Learn More

5-Bromobenzo[b]thiophene 98.0+%, TCI America™

Catalog No. p-7123152
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
B37621G 1 g
B37625G 5 g
2 options

Catalog No. B37621G

Supplier: TCI America B37621G

Only null left

Chemical Identifiers

CAS 4923-87-9
Molecular Formula C8H5BrS
Molecular Weight (g/mol) 213.09
MDL Number MFCD03069318
InChI Key RDSIMGKJEYNNLF-UHFFFAOYSA-N
Synonym 5-bromobenzo b thiophene, 5-bromobenzothiophene, 5-bromo-benzo b thiophene, 5-bromothianaphthene, benzo b thiophene, 5-bromo, zlchem 181, pubchem7752, 5-bromo-benzothiophene, pubchem15353, acmc-1anox
PubChem CID 2776578
IUPAC Name 5-bromo-1-benzothiophene
SMILES BrC1=CC=C2SC=CC2=C1

Specifications

Melting Point 48°C
Boiling Point 107°C
Quantity 1 g
Formula Weight 213.09
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromobenzo[b]thiophene