missing translation for 'onlineSavingsMsg'
Learn More

5-Bromobenzo[b]thiophene, 98+%

Catalog No. p-7048036
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2013303 1 g
AAL2013306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2013303 Supplier Thermo Scientific Chemicals Supplier No. L2013303

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4923-87-9 | C8H5BrS | 213.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4923-87-9
Molecular Formula C8H5BrS
Molecular Weight (g/mol) 213.09
MDL Number MFCD03069318
InChI Key RDSIMGKJEYNNLF-UHFFFAOYSA-N
Synonym 5-bromobenzo b thiophene, 5-bromobenzothiophene, 5-bromo-benzo b thiophene, 5-bromothianaphthene, benzo b thiophene, 5-bromo, zlchem 181, pubchem7752, 5-bromo-benzothiophene, pubchem15353, acmc-1anox
PubChem CID 2776578
IUPAC Name 5-bromo-1-benzothiophene
SMILES BrC1=CC=C2SC=CC2=C1

Specifications

Melting Point 44°C to 48°C
Boiling Point 105°C to 107°C (10 mmHg)
Quantity 1 g
Beilstein 112878
Sensitivity Light sensitive
Formula Weight 213.1
Percent Purity ≥98%
Chemical Name or Material 5-Bromobenzo[b]thiophene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only