missing translation for 'onlineSavingsMsg'
Learn More

5-Bromoacenaphthene 93.0+%, TCI America™

Catalog No. p-7118969
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
B12835G 5 g
B128325G 25 g
2 options

Catalog No. B12835G

Supplier: TCI America B12835G

Only null left

Chemical Identifiers

CAS 2051-98-1
Molecular Formula C12H9Br
Molecular Weight (g/mol) 233.11
MDL Number MFCD00003809
InChI Key QALKJGMGKYKMKE-UHFFFAOYSA-N
Synonym 5-bromoacenaphthene, acenaphthene, 5-bromo, acenaphthylene, 5-bromo-1,2-dihydro, 5-bromoacenaphtene, 5-bromo acenaphthene, akos 51, maybridge1_006415, acmc-1ccr7
PubChem CID 74923
IUPAC Name 5-bromo-1,2-dihydroacenaphthylene
SMILES BrC1=C2C=CC=C3CCC(C=C1)=C23

Specifications

Melting Point 55°C
Color Yellow
Boiling Point 335°C
Quantity 5 g
Formula Weight 233.11
Percent Purity ≥93.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromoacenaphthene