missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-m-xylene, 98%, Thermo Scientific™

Chemical Identifiers

CAS 556-96-7
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.06
InChI Key LMFRTSBQRLSJHC-UHFFFAOYSA-N
Synonym 5-bromo-m-xylene, 3,5-dimethylbromobenzene, benzene, 1-bromo-3,5-dimethyl, m-xylene, 5-bromo, 5-bromo-1,3-xylene, 5-bromo-1,3-dimethylbenzene, 1-bromo-3,5-dimethyl-benzene, 5-bromo-meta-xylene, 5-bromo-3,5-dimethylbenzene, 1-bromo-3,5-dimethyl benzene
PubChem CID 136357
IUPAC Name 1-bromo-3,5-dimethylbenzene
SMILES CC1=CC(=CC(=C1)Br)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC294000250
SDS View Documents
Thermo Scientific Chemicals
294000250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC294001000
SDS View Documents
Thermo Scientific Chemicals
294001000
100g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

556-96-7
185.06
5-bromo-m-xylene, 3,5-dimethylbromobenzene, benzene, 1-bromo-3,5-dimethyl, m-xylene, 5-bromo, 5-bromo-1,3-xylene, 5-bromo-1,3-dimethylbenzene, 1-bromo-3,5-dimethyl-benzene, 5-bromo-meta-xylene, 5-bromo-3,5-dimethylbenzene, 1-bromo-3,5-dimethyl benzene
1-bromo-3,5-dimethylbenzene
C8H9Br
LMFRTSBQRLSJHC-UHFFFAOYSA-N
136357
CC1=CC(=CC(=C1)Br)C

Specifications

556-96-7
87°C
98%
C8H9Br
1.5475 to 1.5495
1.36
LMFRTSBQRLSJHC-UHFFFAOYSA-N
1-bromo-3,5-dimethylbenzene
136357
98%
1.3600g/mL
Authentic
Glass bottle
(CH3)2C6H3Br
25g
5-bromo-m-xylene, 3,5-dimethylbromobenzene, benzene, 1-bromo-3,5-dimethyl, m-xylene, 5-bromo, 5-bromo-1,3-xylene, 5-bromo-1,3-dimethylbenzene, 1-bromo-3,5-dimethyl-benzene, 5-bromo-meta-xylene, 5-bromo-3,5-dimethylbenzene, 1-bromo-3,5-dimethyl benzene
CC1=CC(=CC(=C1)Br)C
185.06
185.06
5-Bromo-m-xylene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not eat,drink or smoke when using this product.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doc

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only