missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-8-hydroxyquinoline 96.0+%, TCI America™

Catalog No. B33245G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33245G 5 g
B332425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B33245G Supplier TCI America Supplier No. B33245G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1198-14-7
Molecular Formula C9H6BrNO
Molecular Weight (g/mol) 224.057
MDL Number MFCD00457011
InChI Key WIIUANWSGSTCLG-UHFFFAOYSA-N
Synonym 5-Bromo-8-quinolinol
PubChem CID 70974
IUPAC Name 5-bromoquinolin-8-ol
SMILES C1=CC2=C(C=CC(=C2N=C1)O)Br

Specifications

Melting Point 127°C
Color White-Yellow
Quantity 5 g
Formula Weight 224.06
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-8-hydroxyquinoline
RTECSNumber VC4582000
TSCA No