missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 5-Bromo-7-chloro-1H-indazole, 96%

Catalog No. AAH3681606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3681606 5 g
AAH3681603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3681606 Supplier Thermo Scientific™ Supplier No. H3681606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 635712-44-6
Molecular Formula C{7}H{4}BrClN{2}
MDL Number MFCD11044588

Specifications

Melting Point ca 205°C dec.
Density 1.787
Boiling Point 364.1°C at760mmHg
Quantity 5 g
Formula Weight 231.48
Percent Purity 96%
Physical Form Solid
ShelfLife 5 Years